Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

solute carrier family 26, member 9, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004768 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-768 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-768 Supplier Abnova Corporation Supplier No. H00115019A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SLC26A9.

This gene is one member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures yet have markedly different tissue expression patterns. The product of this gene is a highly selective chloride ion channel regulated by WNK kinases. Alternative splicing results in multiple transcript variants encoding differing isoforms

Sequence: QTQFRNGYALAQVMDTDIYVNPKTYNRAQDIQGIKIITYCSPLYFANSEIFRQKVIAKTGMDPQKVLLAKQKYLKKQEKRRMRPTQQRRSLFMKTKTVSLQELQQDFENA

Specifications

Antigen solute carrier family 26, member 9
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SLC26A9.
Formulation 50% glycerol
Gene SLC26A9
Gene Accession No. NM_052934
Gene Symbols SLC26A9
Host Species Mouse
Immunogen SLC26A9 (NP_443166, 496 a.a. ∼ 605 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 115019
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.