Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A4 regulator, Mouse, Clone: 4C10, Abnova™

Catalog No. 89001321 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-321 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-321 Supplier Abnova Corporation Supplier No. H00056731M02
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SLC2A4RG.

The protein encoded by this gene is a nuclear transcription factor involved in the activation of the solute carrier family 2 member 4 gene. The encoded protein interacts with another transcription factor, myocyte enhancer factor 2, to activate transcription of this gene. [provided by RefSeq

Sequence: EAAHFLFGEPTLRKRKSPAQVMFQCLWKSCGKVLSTASAMQRHIRLVHLGRQAEPEQSDGEEDFYYTELDVGVDTLTDGLSSLTPVSPTASM

Specifications

Antigen SLC2A4 regulator
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SLC2A4RG.
Formulation PBS with no preservative; pH 7.4
Gene SLC2A4RG
Gene Accession No. NM_020062
Gene Alias GEF/HDBP1/Si-1-2/Si-1-2-19
Gene Symbols SLC2A4RG
Host Species Mouse
Immunogen SLC2A4RG (NP_064446, 178 a.a. ∼ 269 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 56731
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.