Learn More
ABclonal Technology SLC4A7 Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology A10271
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 240-400 of human SLC4A7 (NP_003606.3).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse•Validated Applications:WB•Swiss Prot: Q9Y6M7•Gene ID:9497•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-9962
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.