Learn More
MBL International Corporation Anti-SLC6A2 (NET) (Human) pAb (Polyclonal Antibody)
SDP
Supplier:  MBL International Corporation BMP029
Encompass_Preferred
Clonality:Polyclonal; Application:FCM|ICC|IHC|WB; Conjugate:Unlabeled; Species Reactivity:Human; Size:50 µl; Target:SLC6A2/NET; Polyclonal antibody of 50 µl targeting SLC6A2/NET for FCM, ICC, IHC, WB.SLC6A2/NET Polyclonal Antibody.
Catalog No. 50-453-17
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.