Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SMPD3, Human, IP-WB Antibody Pair, Abnova™

Catalog No. 89301172 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-301-172 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-301-172 Supplier Abnova Corporation Supplier No. H00055512PW1
Add to Cart
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Comprend

Antibody pair for IP: rabbit polyclonal anti-SMPD3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-SMPD3 (50μg)

For immunoprecipitation and to detect the precipitated protein in western blot.

Spécifications

Antigen SMPD3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene SMPD3
Gene Alias FLJ22593/MGC138443/NSMASE2
Gene Symbols SMPD3
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55512
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-SMPD3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-SMPD3 (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Afficher plus de résultats Afficher moins de résultats

For Research Use Only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.