Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006265 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-265 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-265 Supplier Abnova Corporation Supplier No. H00006645B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SNTB2 protein.

Dystrophin is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes. [provided by RefSeq

Sequence: MRVAAATAAAGAGPAMAVWTRATKAGLVELLLRERWVRVVAELSGESLSLTGDAAAAELEPALGPAAAAFNGLPNGGGAGDSLPGSPSRGLGPPSPPAPPRGPAGEAGASPPVRRVRVVKQEAGGLGISIKGGRENRMPILISKIFPGLAADQSRALRLGDAILSVNGTDLRQATHDQAVQALKRAGKEVLLEVKFIREVTPYIKKPSLVSDLPWEGAAPQSPSFSGSEDSGSPKHQNSTKDRKIIPLKMCFAARNLSMPDLENRQN

Specifications

Antigen syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SNTB2 protein.
Formulation No additive
Gene SNTB2
Gene Accession No. ENST00000360496
Gene Alias D16S2531E/EST25263/SNT2B2/SNT3/SNTL
Gene Symbols SNTB2
Host Species Mouse
Immunogen SNTB2 (ENSP00000353686, 1 a.a. ∼ 267 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6645
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.