Learn More
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-SNX1 (100μg); Mouse purified polyclonal anti-SNX1 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
ELISA
Specifications
Specifications
| Antigen | SNX1 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SNX1. |
| Gene | SNX1 |
| Gene Alias | HsT17379/MGC8664/SNX1A/Vps5 |
| Gene Symbols | SNX1 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.