Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin Rat anti-Human, Rat, Rabbit, Clone: YC7, MilliporeSigma™

Catalog No. MAB354MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB354MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB354MI Supplier MilliporeSigma Supplier No. MAB354
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Somatostatin Antibody, clone YC7 is an antibody against Somatostatin for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Somatostatin
Applications Immunohistochemistry
Classification Monoclonal
Clone YC7
Conjugate Unconjugated
Formulation UnPurified tissue culture supernatant containing 0.05% thimerosal.
Gene Accession No. NM_001048.3
Gene Symbols SST, somatostatin, somatostatin-28, somatostatin-14, SMST
Host Species Rat
Immunogen Synthetic peptide corresponding to amino acids 1-14 of cyclic somatostatin conjugated to bovine thyroglobulin using carbodiimide.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.