Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRP54 Mouse, Unlabeled, Clone: 30, BD

Catalog No. BDB610940 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610940 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610940 Supplier BD Biosciences Supplier No. 610940
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ribosomes that exist freely in the cytosol or those attached to the ER are intrinsically the same in their translational properties. ER-bound ribosomes are responsible for the production of secretory proteins and integral ER, Golgi, lysosomal, and plasma membrane spanning proteins. Such proteins contain signal sequences that direct their synthesis to the ER membrane. As the nascent polypeptide emerges from the ribosome, a signal recognition particle (SRP) binds to the signal sequence and serves to couple the ribosome to the protein-translocating machinery in the ER membrane. Although the SRP is a 325kDa ribonucleoprotein, its 54kDa subunit (SRP54) mediates interaction with, and targeting of, the nascent protein to the ER. Via its C-terminal M-domain, SRP54 associates with the nascent protein and inhibits its elongation. This complex binds to the SRP receptor on the ER, the ribosome is delivered to the translocation machinery, SRP is released, and elongation resumes. Targeting and insertion are tightly coupled to a GTPase cycle that involves SRP54 and SRP receptor. Although the mechanisms are unclear, release of SRP from the ER-bound complex requires GTP hydrolysis.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP54
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human SRP54 aa. 262-476
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.