Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-4 Mouse anti-Human, Phycoerythrin, Clone: MC-813-70, MilliporeSigma™

Catalog No. FCMAB116PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB116PMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB116PMI Supplier MilliporeSigma Supplier No. FCMAB116P
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Milli-Mark™ Anti-SSEA-4-PE Antibody, clone MC-813-70 is validated for use in flow cytometry, IC for the detection of SSEA-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA-4
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone MC-813-70
Conjugate PE
Formulation Purified mouse monoclonal IgG3 conjugated to PE fluorochrome in PBS with less than 0.09% sodium azide and 15mg/mL BSA.
Gene Symbols SSEA-4, SSEA4
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep.
Purification Method Protein A purfied
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.