Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat4 (pY693) Mouse, Alexa Fluor 647, Clone: 38/P-STAT4, BD

Catalog No. BDB558137 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558137 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558137 Supplier BD Biosciences Supplier No. 558137
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Stat proteins function both as cytoplasmic signal transducers and as activators of transcription. Seven mammalian Stat proteins have been identified: Stat1-4, Stat5a, 5b, and Stat6. Stat4 has been shown to play an important role in development of T helper cells, specifically the Th1 subset. Stat4 is activated by IL-12 and by type 1 interferons. Knockout mice supported the role that Stat4 plays in IL-12 signaling because lymphocytes from Stat 4-/- mice could not differentiate into Th1 cells or produce IFNγ in response to treatment with IL-12. IFNγ plays an important role in host defense. A key component in the activation of Stat4 is the phosphorylation on tyrosine and serine residues in response to IL-12 stimulation. IL-12 stimulation leads to the phosphorylation of Stat4 on tyrosine 693 and serine 721. Transcriptional activity of Stat4 has been shown to be significantly reduced when residues Y693 and S721 are mutated.

Host Species: Mouse
Clone: 38
Isotype: IgG2b κ
Species Reactivity: Human
Immunogen: Phosphorylated Human Stat4 (pY693)

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat4 (pY693)
Applications Flow Cytometry
Classification Monoclonal
Clone 38/P-STAT4
Conjugate Alexa Fluor 647
Description Signal transducer and activator of transcription 4; SLEB11
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Stat4 (pY693)
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.