Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat5 (pY694) Mouse anti-Human, Unlabeled, Clone: 47, BD

Catalog No. BDB611965 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611965 150 μg
BDB611964 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611965 Supplier BD Biosciences Supplier No. 611965
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Stat proteins function both as cytoplasmic signal transducers and as activators of transcription. Seven mammalian STATs have been identified: Stat 1-4, Stat5a, 5b, and Stat6. The SH3 domain of Stat5 shows extensive homology with the SH2 domains of Stat1 and Stat2. This homology leads to speculation that it may have a broader physiological role than originally anticipated and has led to its classification as a member of the Stat family. Stat5 has been further characterized and shown to be encoded by two separate genes, Stat5a and Stat5b that share over 90% identity at the amino acid level. Further insight into the exact biological role of Stat5a and Stat5b was determined with the generation of Stat5a and Stat5b deficient mice. Stat5a was shown to be involved in lactogenesis and mammary development, while Stat5b was shown to be involved in growth hormone signaling and to play a role in liver gene expression. Both Stat5a and Stat5b share similarities, both are involved in IL-2 induced peripheral T cell proliferation. Stat5 (MGF) mRNA expression is highest in mammary gland tissue. Lower levels are found in ovary, thymus, spleen, kidney, lung, muscle, and the adrenal gland. The peptide hormone, prolactin, binds to the prolactin receptor (PRLR) to initiate the lactogenic response. There are at least three forms of PRLR; however, only the long form is able to activate the 92kDa Stat5 protein by inducing phosphorylation at Tyr694. Once phosphorylated, Stat5 becomes an essential transcription factor which binds to the β-casein gene promoter. Stat5 activity is tightly regulated throughout gestation, lactation, and post-lactation. Treatment of activated Stat5 with a protein tyrosine phosphatase results in the loss of DNA binding activity. The presence of an SH2 domain within Stat5 suggests that it may directly interact with protein tyrosine kinases (PTKs) such as JAK2.

Host Species: Mouse
Clone: 47
Isotype: IgG1, κ
Species Reactivity [for Features Main]: Human
Immunogen: Phosphorylated Sheep Stat5 Peptide

Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat5 (pY694)
Applications Western Blot
Classification Monoclonal
Clone 47
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Sheep Stat5 Peptide
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.