Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Anti-Static Nitrile Finger Cots

Catalog No. 19017535B Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-017-535B Medium
19-017-535C Large
19-017-535D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-017-535B Supplier Honeywell™ North™ Supplier No. 125LNR/MH5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Powder-free finger cots feature a low particulate count, and help to eliminate damaging static discharge in a wide variety of applications.

  • Patented nitrile formulation is comfortable with a high level of stretch
  • Low extractables result in less contamination from reactive ions
  • Helps to eliminate damaging static discharge in a wide variety of applications
  • Free of natural latex proteins that may cause allergic reactions
  • Uniform thickness with excellent tactile sensitivity

Specifications

Certifications/Compliance OSHA Standard - 29 CFR 1915.157
Applications Industrial
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type ESD Finger Cot
Size Medium
Thickness (English) 3 mil
For Use With (Application) Manufacturing Processes
Glove Cuff Style Rolled
Length (English) 2.8 in.
Length (Metric) 7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.