Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD anti-Streptavidin, APC-Cy™7

Catalog No. BDB554063 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554063 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554063 Supplier BD Supplier No. 554063
Add to Cart
Add to Cart

APC conjugate

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature

Host Species:

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptavidin
Concentration 0.2 mg/mL
Conjugate APC-Cy7
Quantity 0.1 mg
Storage Buffer Aqueous buffered solution containing less than or equal to 0.09% sodium azide
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
For Use With (Application) Flow Cytometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.