Learn More
Description
- This antibody is routinely tested by flow cytometric analysis.
- Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Host Species: Flow Cytometry
Specifications
Specifications
| Antigen | Streptavidin |
| Concentration | 0.2 mg/mL |
| Conjugate | PE-Cy5 |
| Quantity | 0.1 mg |
| Regulatory Status | For research use only (RUO) |
| Storage Buffer | Aqueous buffered solution containing less than or equal to 0.09% sodium azide |
| Content And Storage | Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |
| For Use With (Application) | Flow Cytometry |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.