Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ PE-Cy™7 Streptavidin

Catalog No. BDB557598 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557598 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557598 Supplier BD Supplier No. 557598
Add to Cart
Add to Cart

SAv-PE-Cy7 is a useful second-step reagent for the indirect immunofluorescent staining of cells in combination with biotinylated primary antibodies for flow cytometric analysis.

  • Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptavidin
Concentration 0.2 mg/mL
Conjugate PE-Cy7
Quantity 0.1 mg
Regulatory Status For research use only (RUO)
Storage Buffer Aqueous buffered solution containing less than or equal to 0.09% sodium azide
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
For Use With (Application) Flow Cytometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.