Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SUFU, GLI1, Human, Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89301581 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-301-581 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-301-581 Supplier Abnova Corporation Supplier No. DI0625

missing translation for 'validMainframeMsg'

Includes

SUFU rabbit purified polyclonal antibody (20μg) , GLI1 mouse monoclonal antibody (40μg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

For use in in situ Proximity Ligation Assay.

Specifications

Antigen SUFU
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene SUFU
Gene Alias PRO1280/SUFUH/SUFUXL
Gene Symbols SUFU
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51684/2735
Includes Antibody pair set content: 1. SUFU rabbit purified polyclonal antibody (20 ug) 2. GLI1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type SUFU & GLI1 Protein Protein Interaction Antibody Pair
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.