Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Mouse, Clone: 1F8, Abnova™

Catalog No. 89001486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-486 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-486 Supplier Abnova Corporation Supplier No. H00006817M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SULT1A1.

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene. [provided by RefSeq

Sequence: MELIQDTSRPPLEYVKGVPLIKYFAEALGPLQSFQARPDDLLISTYPKSGTTWVSQILDMIYQGGDLEKCHRAPIFMRVPFLEFKAPGIPSGMETLKDTPAPRLLKTHLPLALLPQTLLDQKVKVVYVARNAKDVAVSYYHFYHMAKVHPEPGTWDSFLEKFMVGEVSYGSWYQHVQEWWELSRTHPVLYLFYEDMKENPKREIQKILEFVGHSLPEETVDFVVQHTSFKEMKKNPMTNYTTVPQEFMDHSISPFMRKGMAGDWKTTFTVAQNERFDADYAEKMAGCSLSFRSEL

Specifications

Antigen sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant SULT1A1.
Formulation ascites with no preservative
Gene SULT1A1
Gene Accession No. BC000923
Gene Alias HAST1/HAST2/MGC131921/MGC5163/P-PST/PST/ST1A3/STP/STP1/TSPST1
Gene Symbols SULT1A1
Host Species Mouse
Immunogen SULT1A1 (AAH00923, 1 a.a. ∼ 295 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6817
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.