Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-SV40 T Antigen Clone: PAb416,

Catalog No. DP02200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DP02200UG 200 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DP02200UG Supplier MilliporeSigma™ Supplier No. DP02200UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SV40 T Antigen Clone: PAb416 in samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation

SV40 T Ag (Ab-2) is a mouse monoclonal antibody with specificity for antigenic determinants unique to the SV40 large T antigen and non-reactive with SV40 small T antigen. Both antigens are encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular transforming protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the p105 product of the retinoblastoma susceptibility gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV40 T Antigen
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone PAb416
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.
Host Species Mouse
Immunogen purified SV40 large T-antigen
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes ∽94kDa SV40 large T antigen in SV80 and SV-T2 cells.
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.