Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syk (pY348) Mouse, Alexa Fluor 488, Clone: I120-722, BD

Catalog No. BDB560081 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560081 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560081 Supplier BD Biosciences Supplier No. 560081
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Syk is a non-receptor protein-tyrosine kinase that is closely related to ZAP70 and plays crucial roles in the development and receptor-mediated signaling of most leukocytes and in vascular integrity. Syk is expressed in hematopoietic cells, including B lymphocytes, immature (CD4, CD8 double-negative and double-positive) thymocytes, and myeloid cells, epithelial cell lines, and normal breast tissue. Mature (CD4 or CD8 single-positive) thymocytes and peripheral αβ TCR-bearing T lymphocytes have very low or undetectable levels of Syk. Syk contributes to the signal transduction process by binding to ITAMs ( I mmunoreceptor T yrosine-based A α and Ig β (CD79a and b), TCR ζ , CD3 ε , and FcR γ. Upon receptor activation, Syk binds to phosphorylated ITAMs via its two N-terminal SH2 domains thereby activating Syk and causing tyrosines in the interdomain, between the SH2 and Kinase domains of Syk, to undergo auto-phosphorylation and phosphorylation by Lyn. The tyrosine 348 phosphorylation site (pY348) in human Syk is orthologous to tyrosine 342 in mouse and rat Syk and tyrosine 315 in human ZAP70. This phosphorylated site can act as a binding site for other signaling molecules, such as PLC γ, Vav, and Fgr.

The I120-772 antibody is specific for human Syk (pY348) and does not cross-react with phosphorylated Zap70. The orthologous phosphorylation site in mouse and rat Syk is Y342.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Syk (pY348)
Applications Flow Cytometry
Classification Monoclonal
Clone I120-722
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Syk Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.