Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Mouse, Unlabeled, Clone: 2, BD

Catalog No. BDB611880 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611880 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611880 Supplier BD Biosciences Supplier No. 611880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Synaptophysin is a major integral membrane glycoprotein that was isolated from the presynaptic vesicles of neurons. The structure of synaptophysin includes four transmembrane domains (TMD), an N-temrinal glycosylation site, nine repeats of basic tetrapeptide motifs (GYGP), multiple collagenase sensitive sites (XGP), and several collagen-type tripeptides (GXX). Synaptophysin mRNA is expressed in nervous tissues, as well as many neuroendocrine tumor cell lines. In PC12 cells and brain synaptic vesicles, synaptophysin binds cholesterol, and cholesterol depletion blocks the biogenesis of synaptic-like microvesicles. In addition, synaptophysin binds dynamin in a Ca2+-dependent manner, and injection of peptides that disrupt this interaction decreases neurotransmitter release during high-frequency stimulation of squid neurons. This decrease in neurotransmitter release results from inhibition of synaptophysin-dependent vesicle retrieval. Thus, synaptophysin may have important roles in the biogenesis and recycling of synaptic vesicles. Synaptophysin is an integral membrane glycoprotein with a calculated molecular weight of 33kD, but has been reported to be observed at 38-42kD due to glycosylation.

Host Species: Mouse
Clone: 2
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Synaptophysin aa. 205-306

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptophysin
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Synaptophysin aa. 205-306
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.