Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 4 Mouse anti-Human, Unlabeled, Clone: 49, BD

Catalog No. BDB610439 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610439 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610439 Supplier BD Biosciences Supplier No. 610439
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Signal transmission between neurons is regulated by the release of neurotransmitters at the synapse. This process is controlled by a complex pathway of membrane trafficking in the presynaptic nerve terminal, which leads to membrane fusion and subsequent neurotransmitter secretion. Syntaxin is involved in three important protein complexes that modulate this process: syntaxin and n-sec1, syntaxin, VAMP and SNAP-25, and syntaxin, VAMP, SNAP-25, αSNAP, and NSF (20S complex). A model has been proposed to explain docking, activation, and fusion of synaptic vesicles with donor membranes. This model suggests that VAMP/synaptobrevin and synaptotagmin (vSNARE) on the synaptic vesicle, and SNAP-25 and syntaxin (tSNAREs) on the plasma membrane, interact to form a 7S complex. It appears that syntaxin associates with -sec1 prior to and/or during the formation of the 7S complex. Two additional soluble proteins, αSNAP and NSF, associate with this complex as synaptotagmin releases from the complex. The resulting 20S complex contains syntaxin, SNAP-25, VAMP, αSNAP, and NSF. A large region including the N-terminus is involved in binding n-sec1 while both VAMP and SNAP-25 bind within residues 199-288 of
syntaxin.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 4
Applications Western Blot
Classification Monoclonal
Clone 49
Concentration 250μg/mL
Conjugate Unconjugated
Description STX4
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.