Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Synuclein Mouse, Unlabeled, Clone: 42, BD

Catalog No. p-3851614 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610786 50 μg
BDB610787 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610786 Supplier BD Biosciences Supplier No. 610786
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Synuclein cDNA was isolated from Torpedo californica and found to encode a 143 amino acid neuron-specific protein. In addition, a rat brain cDNA library yielded several clones that encode different proteins with homology to Torpedo Synuclein. A 140 amino acid protein in rat is referred to as Synuclein-1. In situ hybridization shows synuclein mRNAs in discrete areas of rat brain, notably the hippocampus. In humans, two synuclein proteins of 140 and 134 amino acids were sequenced that exhibit a high degree of homology with each other (61% identity) and with previously described Synuclein proteins. The 140 amino acid protein (α-Synuclein) is the human homologue of rat Synuclein-1 and is identical to the non-amyloid-β component precursor (NACP), a presynaptic protein involved in amyloidogenesis in Alzheimer's disease (AD). The 134 amino acid protein (β-Synuclein) is the human homologue of bovine phosphoneuroprotein. These proteins are expressed in brain, primarily in presynaptic nerve terminals. Although the exact function of the Synucleins has not been determined, they have been linked to the prominent neurodegenerative disorders AD and Parkinson's disease.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen α-Synuclein
Applications Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Synuclein-1 aa. 15-123
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.