Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8, CD4 and CD3 cocktail , APC, FITC, PE, Clone: SK1, BD

Catalog No. BDB558625 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558625 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558625 Supplier BD Biosciences Supplier No. 558625
Only null left
Add to Cart
Add to Cart

Monoclonal Antibody

Cocktail Component Clone Isotype

APC anti-Human CD8 SK1 mIgG1

FITC anti-Human CD3 SP34-2 mIgG1

The NHP T Lymphocyte Cocktail is a three-color reagent cocktail designed to identify NHP T lymphocytes by direct immunofluorescence staining with flow cytometric analysis. The SK1 antibody reacts with the hinge-like membrane-proximal domain of the 32-kDa alpha chain of the CD8 differentiation antigen.. The CD8α and β chains (CD8a and CD8b, respectively) form a heterodimer on the surface of most thymocytes and a subpopuplation of mature T-lymphocytes (ie, MHC class I-restricted T cells, including most T suppressor/cytotoxic cells). The L200 antibody reacts with the human form of the 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. L200 antibody also cross-reacts with a subset of CD3-positive peripheral blood lymphocytes, but not monocytes, of both rhesus and cynomolgus macaque monkeys. Cross-reactivity on both lymphocytes and monocytes (weak) of baboon is also observed. The distribution on lymphocytes is similar for both human and monkey, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers. The SP34-2 antibody reacts with the T-cell receptor-associated CD3 cell-surface antigen found on thymocytes and peripheral T lymphocytes. Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8, CD4 and CD3 cocktail
Applications Flow Cytometry
Classification Monoclonal
Clone SK1
Conjugate FITC/APC/PE
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.