Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γδ T Lymphocytes Rat anti-Pig, Unlabeled, Clone: MAC320, BD

Catalog No. BDB551543 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551543 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551543 Supplier BD Biosciences Supplier No. 551543
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The MAC320 antibody monoclonal antibody specifically binds to the 270- and 280-kDa isoforms of the SWC6 heterodimer expressed on Null T cells. These cells form a unique population of CD2-, surface Ig-, and thymus-dependent nylon wool-nonadherent peripheral blood lymphocytes found at high frequencies in young pigs. Null T cells are also CD4- and CD8-, and the majority of them express γδ T-cell receptors (TCR). In addition to peripheral blood, MAC320+ leukocytes (CD2- γδ T cells) are found in the thymus, especially in the medulla and in the marginal zone and red pulp of the spleen. They are rare in normal lymph nodes, tonsils, Peyer's patches, bone marrow, and skin. An additional population of MAC320- CD2+ γδ T cells is found in the blood and spleen. The MAC320 antibody does not react with pig erythrocytes, monocytes, or granulocytes, nor with cow, sheep, or human lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen γδ T Lymphocytes
Applications Flow Cytometry
Classification Monoclonal
Clone MAC320
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Null T cells purified from the blood of young pigs
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.