Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAP1 Mouse anti-Human, Clone: mAb 148.3, MilliporeSigma™

Catalog No. MABF125MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF125MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF125MI Supplier MilliporeSigma Supplier No. MABF125
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Antigen peptide transporter 1 using this mouse monoclonal antibody, Anti-TAP1 Antibody, clone mAb 148.3 validated for use in western blotting, immunoprecipitation, Immunofluorescence and Activity Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP1
Applications Functional Assay, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mAb 148.3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ supernatant in buffer containing PBS without preservatives.
Gene Accession No. NP_000584
Gene Symbols TAP1, ABCB2, PSF1, RING4, Y3
Host Species Mouse
Immunogen Linear peptide corresponding to the CYWAMVQAPADAPE sequence of human TAP1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.