Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tartrate Resistant Acid Phosphatase Monoclonal Antibody (26E5), Invitrogen™

Catalog No. PIMA512387
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512387 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512387 Supplier Thermo Scientific Supplier No. MA512387

Mouse Monoclonal Antibody

MA5-12387 targets Tartrate Resistant Acid Phosphatase in IHC (P) applications and shows reactivity with Human samples. The MA5-12387 immunogen is recombinant protein corresponding to N-terminal region of human TRAP.

Tartrate Resistant Acid Phosphatase Monoclonal antibody specifically detects Tartrate Resistant Acid Phosphatase in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tartrate Resistant Acid Phosphatase
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 26E5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene ACP5
Gene Accession No. P13686
Gene Alias acid phosphatase 5, tartrate resistant; acid phosphatase type V; Acp5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; TRAP; TR-AP; TRAPase; TrATPase; TTRRAP; Type 5 acid phosphatase
Gene Symbols ACP5
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminal region of human TRAP
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.