Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TATA-box-binding protein Mouse anti-Human, Mouse, Rat, Clone: SL30-3-563, MilliporeSigma™

Catalog No. 051531MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-153-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-153-1MI Supplier MilliporeSigma Supplier No. 051531
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-TATA-box-binding protein Antibody, clone SL30-3-563 (mouse monoclonal antibody) validated in western blotting to detect TATA-box-binding protein also known as TATA box binding protein, Transcription initiation factor TFIID TBP subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TATA-box-binding protein
Applications Western Blot
Classification Monoclonal
Clone SL30-3-563
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_003185
Gene Symbols GTF2D, TFIID, TBP, GTF2D, SCA17, TF2D
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human TATA-box-binding protein at the N-terminus.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.