Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau-1 Mouse anti-Bovine, Human, Rat, Clone: PC1C6, MilliporeSigma™

Catalog No. MAB3420MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3420MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3420MI Supplier MilliporeSigma Supplier No. MAB3420
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tau-1 Antibody, clone PC1C6 is an antibody against Tau-1 for use in immunohistochemistry and western blotting with more than 65 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau-1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone PC1C6
Conjugate Unconjugated
Formulation 0.02M phosphate buffer, pH 7.6, 0.25M NaCl, and 0.1% sodium azide
Gene Accession No. NM_005910.3; NM_016834.2; NM_016835.2; NM_016841.2
Gene Symbols MAPT, MTBT2, MAPTL, tau, FTDP-17, MSTD, TAU, FLJ31424, MTBT1, PHF-tau, DDPAC, MGC138549, PPND
Host Species Mouse
Immunogen Purified denatured bovine microtubule associated proteins.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.