Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau (4-repeat isoform RD4) Mouse anti-Human, Rat, Bovine, Mouse, Clone: 1E1/A6, MilliporeSigma™

Catalog No. 05804MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-804-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-804-MI Supplier MilliporeSigma Supplier No. 05804
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6 is validated for use in immunohistochemistry, western blotting for the detection of Tau (4-repeat isoform RD4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau (4-repeat isoform RD4)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1E1/A6
Conjugate Unconjugated
Formulation Mouse monoclonal cultured supernatant containing 0.05% sodium azide.
Frozen at −20°C.
Gene Accession No. NM_016835
Gene Symbols MAPT, MTBT2, tau, FTDP-17, MSTD, TAU, MTBT1, PHF-tau, MGC138549, MAPTL, FLJ31424, DDPAC, PPND
Host Species Mouse
Immunogen Bovine thyroglobulin-conjugated synthetic peptide corresponding to amino acids 275-291 (VQIINKKLDLSNVQSKC) of human Tau (4-repeat isoform RD4). Region is flanking junction coded by adjacent exons 9 & 11 with the inclusion of exon 10. The immunizing sequ
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.