Learn More
Tau phospho Serine 199/202 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Tau phospho Serine 199/202 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in Dulbecco's PBS (without Mg2+ and Ca2+)), pH 7.3, 50% glycerol with 1.0mg/mL BSA and 0.05% sodium azide. |
| Gene Symbols | MAPT; MTBT2; MAPTL; PHF-tau; tau; DDPAC; FTDP-17; MGC138549; MSTD; TAU; FLJ31424; MTBT1; PPND |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide of amino acids surrounding the phosphoSerine 199 and 202 sites of human Tau. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.