Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCP-1 epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522069
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522069 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522069 Supplier Invitrogen™ Supplier No. PA522069
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Mouse brain. Predicted reactivity: Mouse (95%), Rat (96%), Zebrafish (87%), Xenopus laevis (89%), Pig (97%), Chicken (90%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCP-1 epsilon
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CCT5
Gene Accession No. P48643, P80316
Gene Alias CCT5; CCTE; CCT-epsilon; chaperonin containing TCP1 subunit 5; chaperonin containing Tcp1, subunit 5 (epsilon); chaperonin subunit 5 (epsilon); epididymis secretory protein Li 69; HEL-S-69; Kiaa0098; mKIAA0098; PNAS-102; T-complex protein 1 subunit epsilon; TCP-1 epsilon; TCP-1-epsilon; TCPE; unm tf212; unm_tf212; wu:fb54h08
Gene Symbols CCT5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TCP1 epsilon. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12465, 22948
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.