Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TER-119/Erythroid Cells Rat anti-Mouse, Unlabeled, Clone: TER-119, BD

Catalog No. BDB550565 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550565 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550565 Supplier BD Biosciences Supplier No. 550565
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen TER-119/Erythroid Cells
Applications Flow Cytometry
Classification Monoclonal
Clone TER-119
Concentration 31.25μg/mL
Conjugate Unconjugated
Description Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Fetal Liver
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.