Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

testis expressed 13A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018535 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-535 Supplier Abnova Corporation Supplier No. H00056157D01P
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TEX13A protein.

This gene is similar to a mouse gene that is expressed in the testis. [provided by RefSeq

Sequence: MALRPEDPSSGFRHSNVVAFINEKMARHTKGPEFYLENISLSWEKVEDKLRAILEDSEVPSEVKEACTWGSLALGVRFAHRQAQLQRHRVRWLHGFAKLHKSAAQALASDLKKLREQQETERKEAASRLRMAQTSLVEVQKERDKELVSPHEWEQGAGWPGLATAGGVCTEGAAEEEEEAAVAAAGAAGGKGAEEEQRDVEVVAAPVEAMAPPVEAGAAPMETQFPHVEARAASMETTEKLERILLQLLGDADQEKYTYWGQKEGDLRSVETATSYFSGTTNPWSRASSEPLPVQLPASYSYSYSSPFSSFSDIPTISPPQATVTAPVPPQLPSDWEAFDTSLWSDGGPHRIDHQEHPRDRRYSEPHQQRPPVYRRPGDWDCPWCNAVNFSRRDTCFDCGKGIWLQKPH

Specifications

Antigen testis expressed 13A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TEX13A protein.
Formulation PBS with no preservative; pH 7.4
Gene TEX13A
Gene Accession No. NM_031274
Gene Alias MGC131984
Gene Symbols TEX13A
Host Species Rabbit
Immunogen TEX13A (NP_112564.1, 1 a.a. ∼ 409 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56157
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.