Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TIGIT, APC, Clone: 1G9,

Catalog No. mabf1417mi Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1417MI 100 μg
MABF1416MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1417MI Supplier MilliporeSigma™ Supplier No. MABF1417
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TIGIT Clone: 1G9 in Mouse samples, and it is validated for Flow Cytometry

The 1G9 antibody reacts with mouse TIGIT (T cell Ig and ITIM domain), a 26kDa member of the CD28 receptor family which is reported to regulate T cell receptor (TCR) activation. Within the CD28 family of receptors there are those which have co-stimulatory activity, such as CD28 and CTLA-4, as well as more recently identified receptors like TIGIT which are proposed to provide co-inhibitory signals. TIGIT is expressed and upregulated on activated T cells, and is also expressed on memory and regulatory T cells. Upon engagement by its ligands, CD112 and CD155, TIGIT signaling inhibits T cell proliferation and suppresses T cell responses, without triggering cell deletion. A second inhibitory effect of TIGIT signaling is the generation of immunoregulatory dendritic cells, which secrete IL-10 and TGF-beta to further inhibit T cell function.The 1G9 antibody may be used for flow cytometric analysis of TIGIT, which is expressed at very high levels on T regulatory cells (Tregs) and activated conventional T cells, as well as memory T cells and NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIGIT
Applications Flow Cytometry
Classification Monoclonal
Clone 1G9
Conjugate APC
Formulation Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P86176
Gene Symbols Tigit
Host Species Mouse
Immunogen Proprietary immunogen
Purification Method Purified by size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.