Learn More
Apexbio Technology LLC Anti-TNFRSF10B/TRAILR2/CD262 Antibody (Tigatuzumab) 1mg
SDP
Supplier:  Apexbio Technology LLC F10441
Encompass
Tigatuzumab (CAS 918127-53-4) is a recombinant human IgG1 monoclonal antibody targeting tumor necrosis factor receptor superfamily member 10B (TNFRSF10B) also known as TRAIL receptor 2 (TRAILR2) or CD262 It is designed to bind DR5 a receptor expressed on the surface of diverse cancer cells thereby modulating apoptosis-associated signaling pathways Tigatuzumab exerts its biological activity primarily through induction of apoptosis via receptor-mediated caspase-dependent mechanisms leading to tumor cell death and inhibition of tumor growth Based on these pharmacological properties tigatuzumab holds research potential in oncology and apoptosis-associated signaling pathway studies both in vitro and in vivo
Catalog No. 50-000-94544
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.