Learn More
Description
Includes
Capture antibody: mouse monoclonal anti-TNFSF15 (100μg); Detection antibody: biotinylated mouse monoclonal anti-TNFSF15, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF15.
Host Species: Mouse, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
Specifications
| Antigen | TNFSF15 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF15. |
| Gene | TNFSF15 |
| Gene Alias | MGC129934/MGC129935/TL1/TL1A/VEGI/VEGI192A |
| Gene Symbols | TNFSF15 |
| Host Species | Mouse |
| Quantity | 1 Set |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.