Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-TNP, Alexa Fluor 488, Clone: G155-228, BD
Supplier: BD Biosciences 562409
Description
Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ
Species Reactivity: Rat
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control

Specifications
TNP | |
G155-228 | |
Alexa Fluor 488 | |
TNP-keyhole limpet hemocyanin | |
0.1 mg | |
Primary | |
IgM κ |
Monoclonal | |
0.2mg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction