Learn More
Description
Host Species: Hamster
Clone: A19-3
Isotype: IgG1, κ
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Specifications
| Antigen | TNP |
| Classification | Monoclonal |
| Clone | A19-3 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-CF594 |
| Host Species | Hamster |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.