Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-2-49 Mouse anti-Human, Pig, Rabbit, Feline, Alkaline Phosphatase, Clone: TRA-2-49/6E, MilliporeSigma™

Catalog No. MAB4349MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4349MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4349MI Supplier MilliporeSigma Supplier No. MAB4349
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This Anti-TRA-2-49 Antibody, Liver/Bone/Kidney Alkaline Phosphatase, clone TRA-2-49/6E is validated for use in flow cytometry, immunofluoresence, immunocytochemistry, immunoprecipitation for the detection of TRA-2-49.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-2-49
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone TRA-2-49/6E
Conjugate Alkaline Phosphatase (AP)
Formulation Purified mouse monoclonal IgG1 liquid in buffer containing 0.02MPBS, 0.25MNaCl pH 7.6 with 0.1% Sodium Azide
Gene Symbols TRA-2-49
Host Species Mouse
Immunogen 2102Ep human Embryonal Carcinoma Cells
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Feline, Human, Pig, Rabbit
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.