Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRADD Mouse, Unlabeled, Clone: 37, BD

Catalog No. BDB610572 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610572 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610572 Supplier BD Biosciences Supplier No. 610572

Mouse Monoclonal Antibody

TNFα (Tumor Necrosis Factor) stimulates programmed cell death and NF-κB activation as a result of its binding to the TNF receptor 1 (TNFR1). Within this receptor, a sequence referred to as the “death domain” has been shown to be necessary for both of these functions. Using the yeast two-hybrid system to detect proteins which interact with the receptor through this “death domain”, a 34kDa protein was found and designated TRADD (TNFR1-Associated Death Domain protein). TRADD appears to contain no intrinsic catalytic activity. It also contains a death domain and it has been shown to bind to FADD and RIP. Mutational analysis of TRADD demonstrates that programmed cell death and NF-κB activation are distinct and controlled independently.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRADD
Applications Western Blot
Classification Monoclonal
Clone 37
Concentration 250μg/mL
Conjugate Unconjugated
Description TNFR1-Associated Death Domain protein
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.