Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the quantitative Determination of Anti-Trastuzumab antibodies in human serum and plasma. Assay Range: 40 - 640 ng/ml; Sensitivity: 36 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBI2017
Encompass_Preferred
Pack Size: 1 x 96 wells. Introduction: Trastuzumab is a humanized monoclonal antibody which targets Her-2/neu receptor. It binds to an extracellular domain of this receptor and inhibits HER2 homodimerization, thereby preventing HER2-mediated signaling. ADA has been observed in pre-clinical and clinical studies, resulting in significant changes in toxicology, pharmacokinetics and efficacy. These effects result from the generation of drug-induced (neutralizing) autoantibodies against Trastuzumab and can be responsible for allergic reaction, or even anaphylactic shock. This Anti-Trastuzumab ELISA kit detects Anti-Trastuzumab antibodies and may be used for monitoring immunogenicity. Validated against ICH M10 guidelines for bioassays. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <10%.
Catalog No. NC2747141
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.