Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TREM1 Matched Antibody Pair, Abnova

Catalog No. 89017428 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-428 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-428 Supplier Abnova Corporation Supplier No. H00054210AP21
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TREM1 (100μg)
  • 2. Detection antibody: mouse purified polyclonal anti-TREM1 (20μg)
  • Sufficient for at least 3-5 x 96 well plates using recommended protocols.

Monocyte/macrophage- and neutrophil-mediated inflammatory responses can be stimulated through a variety of receptors, including G protein-linked 7-transmembrane receptors (e.g., FPR1; MIM 136537), Fc receptors (see MIM 146790), CD14 (MIM 158120) and Toll-like receptors (e.g., TLR4; MIM 603030), and cytokine receptors (e.g., IFNGR1; MIM 107470). Engagement of these receptors can also prime myeloid cells to respond to other stimuli. Myeloid cells express receptors belonging to the Ig superfamily, such as TREM1, or to the C-type lectin superfamily. Depending on their transmembrane and cytoplasmic sequence structure, these receptors have either activating (e.g., KIR2DS1; MIM 604952) or inhibitory functions (e.g., KIR2DL1; MIM 604936).

  • Species Reactivity: Human

ELISA

Specifications

Antigen TREM1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TREM1.
Gene TREM1
Gene Alias TREM-1
Gene Symbols TREM1
Host Species Mouse, Rabbit
Immunogen NA; TREM1 (NP_061113.1, 1 a.a. ∽ 234 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54210
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TREM1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-TREM1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.