Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-trimethyl Histone H3 (Lys36) Ab 1, Polyclonal,

Catalog No. p-5168797 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-549-SMI 10 μL
07-549-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-549-SMI Supplier MilliporeSigma™ Supplier No. 07549S

Specifications

Antigen trimethyl Histone H3 (Lys36)
Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum IgG in buffer containing 0.05% sodium azide and 30% glycerol. Liquid at −20°C.
Gene Symbols H3F3A; MGC87783; H3.3A; MGC87782; H3F3; H3.3B; H3F3B
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence...GVme3KKP...in which me3K corresponds to trimethyl lysine 36 of human histone H3.
Purification Method Unpurified
Quantity 10 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.