Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl Histone H3 (Lys4) Mouse anti-Human, Vertebrates, Clone: CMA304, MilliporeSigma™

Catalog No. p-5178501 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-133-9SMI 10 μg
05-133-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-133-9SMI Supplier MilliporeSigma Supplier No. 051339S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Trimethyl Histone H3 (Lys4) Antibody, clone CMA304 is a mouse monoclonal antibody for detection of Trimethyl Histone H3 (Lys4) also known as H3K4me3, Histone H3 (tri methyl K4) and has been validated in ELISA, immunocytochemistry, immunoprecipitation, western blot, Mplex, DB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trimethyl Histone H3 (Lys4)
Applications Dot Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CMA304
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.
Gene Accession No. NM_003493
Gene Symbols H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, trimethylated on Lys4, conjugated to KLH.
Purification Method Protein G purfied
Quantity 10 μg
Primary or Secondary Primary
Target Species Human, Vertebrates
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.