Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Rat, Chicken, Polyclonal, MilliporeSigma™

Catalog No. 07442MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-442-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-442-MI Supplier MilliporeSigma Supplier No. 07442
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, immunocytochemistry, Mplex, PIA, western blot, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).
TRUSTED_SUSTAINABILITY

Specifications

Antigen trimethyl Histone H3 (Lys9)
Applications Multiplex, Dot Blot, Immunocytochemistry, Immunoassay, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
Gene Symbols HIST3H3; H3FT; H3/t; H3t; MGC126888; H3/g; H3.4; H3T; MGC126886
Host Species Rabbit
Immunogen KLH-conjugated, 2X-branched synthetic peptide containing the sequence ...AR[me3K]ST... in which me3K corresponds to trimethyl-lysine at residue 9 of human histone H3.
Purification Method Protein A chromatography
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Chicken, Human, Mouse, Rat
Content And Storage −20°C for 12 months. Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.