Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trop2 Mouse anti-Human, Clone: 2G10, MilliporeSigma™

Catalog No. MABC519MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC519MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC519MI Supplier MilliporeSigma Supplier No. MABC519
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Trop2 antibody, clone 2G10, Azide Free is validated for use in immunocytochemistry, flow cytometry and inhibition for the detection of Trop2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trop2
Applications Flow Cytometry, Immunocytochemistry, Inhibition Assays
Classification Monoclonal
Clone 2G10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives.
Gene Accession No. NP_002344
Gene Symbols TACSTD2, GA733-1, M1S1, TROP2
Host Species Mouse
Immunogen Recombinant protein corresponding to the extracellular domain of human Trop2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.