Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSEN2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017172 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-172 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-172 Supplier Abnova Corporation Supplier No. H00080746B01P
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TSEN2 protein.

This gene encodes one of the subunits of the tRNA splicing endonuclease. This endonuclease catalyzes the first step in RNA splicing which is the removal of introns. Mutations in this gene have been associated with pontocerebellar hypoplasia type 2. A pseudogene has been identified on chromosome 4. Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: MAEAVFHAPKRKRRVYETYESPLPIPFGQDHGPLKEFKIFRAEMINNNVIVRNAEDIEQLYGKGYFGKGILSRSRPSFTISDPKLVAKWKDMKTNMPIITSKRYQHSVEWAAELMRRQGQDESTVRRILKDYTKPLEHPPVKRNEEAQVHDKLNSGMVSNMEGTAGGERPSVVNGDSGKSGGVGDPREPLGCLQEGSGCHPTTESFEKSVREDASPLPHVCCCKQDALILQRGLHHEDGSQHIGLLHPGDRGPDHEYVLVEEAECAMSEREAAPNEELVQRNRLICRRNPYRIFEYLQLSLEEAFFLVYALGCLSIYYEKEPLTIVKLWKAFTVVQPTFRTTYMAYHYFRSKGWVPKVGLKYGTDLLLYRKGPPFYHASYSVIIELVDDHFEGSLRRPLSWKSLAALSRVSVNVSKELMLCYLIKPSTMTDKEMESPECMKRIKVQEVILSRWVSSRERSDQDDL

Specifications

Antigen TSEN2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TSEN2 protein.
Formulation PBS with no preservative; pH 7.4
Gene TSEN2
Gene Accession No. NM_025265.2
Gene Alias MGC2776/MGC4440/PCH2B/SEN2/SEN2L
Gene Symbols TSEN2
Host Species Mouse
Immunogen TSEN2 (NP_079541.1, 1 a.a. ∼ 465 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80746
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.