Learn More
Alpco Ltd Anti Tissue Transglutaminase-G ELISA (Anti-tTg-G ELISA)
SDP
Supplier:  Alpco Ltd 35TGGHUE01
Encompass_Preferred
The Anti Tissue Transglutaminase-G ELISA is a solid phase enzyme immunoassay for the quantitative and qualitative detection of IgG antibodies against neo-epitopes of tissue transglutaminase (tTG) in human serum. The assay employing human recombinant transglutaminase crosslinked with gliadin-specific peptides displays neo-epitopes of tTG which ensures a significantly increased sensitivity and specificity of the test. The assay is a tool for the diagnosis of celiac disease (gluten-sensitive enteropathy). FDA Registered. For In Vitro Diagnostic Use.
Catalog No. NC2329171
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.