Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tumor Necrosis Factor-α Mouse anti-Human, Clone: 195

Catalog No. MAB1096MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1096MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1096MI Supplier MilliporeSigma™ Supplier No. MAB1096
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Tumor Necrosis Factor-α Antibody, clone 195 is validated for use in immunocytochemistry, immunohistochemistry, NEUT, western blotting for the detection of Tumor Necrosis Factor-α.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tumor Necrosis Factor-α
Applications Immunocytochemistry, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 195
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.01M PBS pH 7.1, 0.15M NaCl containing no preservatives. Sterile filtered.
Gene Accession No. NM_000594.2
Gene Symbols TNF, OTTHUMP00000037669, TNFA, TNFSF2, cachectin, TNF-a, Cachectin, DIF, TNF-alpha
Host Species Mouse
Immunogen Human, recombinant tumor necrosis factor-alpha produced in E. coli.
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.